Learn More
Sino Biological Recombinant Anti-IGF1R Antibody (PE), Rabbit Monoclonal
Supplier:  Sino Biological 10164R012P25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IGF1R / CD221 (rh IGF1R / CD221; Catalog#10164-H08H; NP_000866.1; Met1-Asn932) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-172-2259