Learn More
Sino Biological Anti-CD25/IL-2R alpha Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10165MM17F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL2RA/IL-2RA/CD25 (rh IL2RA/IL-2RA/CD25; Catalog#10165-H08H; NP_000408.1; Met1-Cys213) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2282