Learn More
Sino Biological Recombinant Anti-CD25/IL-2R alpha Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10165R216F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD25 / IL-2RA (rh CD25 / IL-2RA; Catalog#10165-H08H; NP_000408.1; Met 1-Cys213) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2293