Learn More
Sino Biological Recombinant Anti-CD25/IL-2R alpha Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10165R702100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD25 / IL2RA (rh CD25 / IL2RA; Catalog#10165-H08H; NP_000408.1; Met1-Cys213).
Catalog No. 50-172-2296