Learn More
Sino Biological Anti-CD25/IL-2R alpha Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10165RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human IL2Ra / CD25 (rh IL2Ra; Catalog#10165-H08H; NP_000408.1; Met 1-Cys 213). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2298