Learn More
Sino Biological Recombinant Human Cystatin F/CST7 Protein (hFc Tag)
Supplier:  Sino Biological 10167H02H20
Encompass_Preferred
A DNA sequence encoding the human Cystatin F precursor (NP_003641.3) (Met 1-His 145) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4190