Learn More
Sino Biological Recombinant Human Cystatin F/CST7 Protein
Supplier:  Sino Biological 10167HCCH20
Encompass_Preferred
The mature form of the human Cystatin F (NP_003641.3) (Met 1-His 145) with five amino acids (DDDDK) at the C-terminus was expressed and purified.
Catalog No. 50-161-4194