Learn More
Sino Biological Anti-Cystatin F/CST7 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10167RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CST7 (rh CST7; Catalog#10167-H08H; NP_003641.3; Met 1-His 145). CST7 specific IgG was purified by human CST7 affinity chromatography.
Catalog No. 50-172-2346