Learn More
Sino Biological Anti-NBL1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10169MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human NBL1 / DAN (rh NBL1; Catalog#10169-H08H; NP_005371.1; Met 1-Asp 180). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2377