Learn More
Sino Biological Recombinant Human DKK-1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10170H08H100
Encompass_Preferred
A DNA sequence encoding the human DKK1 precursor (NP_036374.1) (Met 2-His 266) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4201