Learn More
Sino Biological Recombinant Anti-DKK-1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10170R004100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human dickkopf related protein 1 (rh Dkk1; Catalog#10170-H08H; Met 2-His 266; NP_036374.1).
Catalog No. 50-172-2413