Learn More
Sino Biological Anti-DKK-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10170RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human dickkopf related protein 1 (rhDkk1; Catalog#10170-H08H; NP_036374.1; Met 2-His266). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2417