Learn More
Sino Biological Recombinant Human DLL4 Protein, HPLC-verified
Supplier:  Sino Biological 10171HCCH200
Encompass_Preferred
A DNA sequence encoding the human DLL4 (NP_061947.1) (Met1-Pro524) was expressed with five amino acids (DDDDK) at the C-terminus.
Catalog No. 50-161-4208