Learn More
Sino Biological Anti-DLL4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10171RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human DLL4 / Delta4 extracellular domain (rh DLL4; Catalog#10171-H08H; NP_061947.1; Met 1-Pro 524). DLL4 specific IgG was purified by human DLL4 affinity chromatography.
Catalog No. 50-172-2448