Learn More
Sino Biological Anti-COMP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10173RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human COMP (rhCOMP; Catalog#10173-H08H; NP_000086.2; Met 1-Ala 757). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2472