Learn More
Sino Biological Anti-Contactin 3 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10174MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Contactin-3 (rh CNTN3; Catalog#10174-H08H; Met1-Ser1002,708Asp/Asn; NP_065923.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2490