Learn More
Sino Biological Anti-Contactin 3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10174RP03100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CNTN3 (rh CNTN3; Catalog#10174-H08H; Met1-Ser1002,708Asp/Asn; NP_065923.1). CNTN3 specific IgG was purified by human CNTN3 affinity chromatography.
Catalog No. 50-172-2498