Learn More
Sino Biological Recombinant Human DR6/TNFRSF21 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10175H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Leu 350) of human DR6 (NP_055267.1) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4218