Learn More
Sino Biological Anti-DR6/TNFRSF21 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10175MM03100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human TNFRSF21 / DR6 (rh TNFRSF21 / DR6; Catalog#10175-H08H; NP_055267.1; Met1-Leu350). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2510