Learn More
Sino Biological Recombinant Human IL-18R beta / IL-18RAP Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10176H08H100
Encompass_Preferred
A DNA sequence encoding the human IL18RAP (NP_003844.1) (Met 1-Gly 357) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4224