Learn More
Sino Biological Anti-IL1RAPL1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10177MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-1R8 / IL1RAPL1 (rh IL-1R8 / IL1RAPL1; Catalog#10177-H08H; NP_055086.1; Met 1-Leu 354). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2558