Learn More
Sino Biological Recombinant Human S100A1 Protein (hFc Tag)
Supplier:  Sino Biological 10179H01H100
Encompass_Preferred
A DNA sequence encoding the human S100A1 (NP_006262.1) (Gly 2-Ser94) was expressed with the N-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4232