Learn More
Sino Biological Recombinant Human S100A2 Protein
Supplier:  Sino Biological 10180HNAE100
Encompass_Preferred
A DNA sequence encoding the human S100A2 (NP_005969.1) (Met 2-Pro 98) was expressed.
Catalog No. 50-161-4238