Learn More
Sino Biological Recombinant Human Prokineticin 1/EG-VEGF Protein (His Tag)
Supplier:  Sino Biological 10183H08B50
Encompass_Preferred
A DNA sequence encoding the human EG-VEGF (Met 1-Phe105) (P58294) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-4245