Learn More
Sino Biological Recombinant Human CD147/Basigin/EMMPRIN Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10186H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-His 205) of human CD147 (NP_940991.1) precursor was expressed with the fused polyhistidine tag at the C-terminus.
Catalog No. 50-161-4252