Learn More
Sino Biological Anti-CD147/Basigin/EMMPRIN Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10186MM08F100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD147 / EMMPRIN / Basigin (rh CD147 / EMMPRIN / Basigin; Catalog#10186-H08H; NP_940991.1; Met 1-His 205) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2638