Learn More
Sino Biological Recombinant Anti-CD147/Basigin/EMMPRIN Antibody (APC), Rabbit Monoclonal
Supplier:  Sino Biological 10186R125A100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human WFDC2 / HE4 (rh WFDC2 / HE4; Catalog#12609-H08H; NP_940991.1; Met1-His205) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-2647