Learn More
Sino Biological Recombinant Anti-CD147/Basigin/EMMPRIN Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10186R125F100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD147 / Basigin (rh CD147; Catalog#10186-H08H; NP_940991.1; Met 1-His 205) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2649