Learn More
Sino Biological Anti-CD147/Basigin/EMMPRIN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10186RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD147. (rhCD147; Catalog#10186-H08H; NP_940991.1; Met 1-His 205). CD147 specific IgG was purified by human CD147 affinity chromatography.
Catalog No. 50-172-2656