Learn More
Sino Biological Recombinant Anti-ESAM Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10187R113F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human ESAM (rh ESAM; Catalog#10187-H08H; NP_620411.2; Met 1-Ala 248) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-172-2667