Learn More
Sino Biological Anti-ESAM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10187RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ESAM extracellular domain (rh ESAM; Catalog#10187-H08H; NP_620411.2; Met 1-Ala 248). ESAM specific IgG was purified by human ESAM affinity chromatography.
Catalog No. 50-172-2675