Learn More
Sino Biological Recombinant Human Ephrin A3 Protein (hFc Tag)
Supplier:  Sino Biological 10188H02H100
Encompass_Preferred
A DNA sequence encoding the human EphrinA3 (NP_004943.1) (Met 1-Ser 213) with the C-terminal propeptide removed was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4262