Learn More
Sino Biological Anti-Ephrin A3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10188RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Ephrin-A3 (rh EphrinA3; Catalog#10188-H08H; NP_004943.1; Met 1-Ser 213). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2694