Learn More
Sino Biological Anti-GSTP1/GST3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101887T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human GSTP1, and purified by antigen affinity chromatography.
Catalog No. 50-172-2685