Learn More
Sino Biological Recombinant Human Decorin Protein (hFc Tag)
Supplier:  Sino Biological 10189H01H100
Encompass_Preferred
A DNA sequence encoding the mature form of human Decorin (NP_001911.1) (Asp 31-Lys 359) was expressed with the fused Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-4271