Learn More
Sino Biological Recombinant Human Decorin Protein (His Tag)
Supplier:  Sino Biological 10189H08H100
Encompass_Preferred
A DNA sequence encoding the human Decorin (NP_001911.1) (Met 1-Lys 359) was expressed, with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4273