Learn More
Sino Biological Anti-Decorin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10189RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Decorin (rh Decorin; Catalog#10189-H08H; NP_001911.1; Met 1-Lys 359). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2705