Learn More
Sino Biological Recombinant Human Ephrin A5 Protein (hFc Tag)
Supplier:  Sino Biological 10192H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (NP_001953.1)(Met 1-Asn 203) of human Ephrin-A5 (NM_001962.1) precursor was expressed with the C-terminal fused Fc region of human IgG1.
Catalog No. 50-161-4277