Learn More
Sino Biological Recombinant Human Ephrin A5 Protein (His Tag)
Supplier:  Sino Biological 10192H08H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Asn 203) of human Ephrin-A5 (NP_001953.1) precursor was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4279