Learn More
Sino Biological Anti-LY75/CD205 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101920T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse LY75 / DEC-205 / CD205, and purified by antigen affinity chromatography.
Catalog No. 50-172-2720