Learn More
Sino Biological Anti-Bcl-2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10195MM17100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Bcl-2 (rh Bcl-2; Catalog#10195-H08E; P10415-1; Met1-Asp211). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2770