Learn More
Sino Biological Anti-Bcl-2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10195T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human BCL2 / Bcl-2 (rh BCL2 / Bcl-2; Catalog#10195-H08E; P10415-1; Met1-Asp211). BCL2 / Bcl-2 specific IgG was purified by Human BCL2 / Bcl-2 affinity chromatography.
Catalog No. 50-172-2773