Learn More
Sino Biological Recombinant Human EphB6 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 10197H02H100
Encompass_Preferred
The extracellular domain (Met 1-Ser 579) of human EphB6 (NP_004436.1) precursor was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4286