Learn More
Sino Biological Anti-EphB6 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10197MM07100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human EphB6 (rh EphB6; Catalog#10197-H08H; NP_004436.1; Met 1-Ser 579). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2791