Learn More
Sino Biological Anti-EphB6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10197T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human EphB6 / Eph Receptor B6 (rh EphB6 / Eph Receptor B6; Catalog#10197-H08H; NP_004436.1; Met1-Ser579). EphB6 / Eph Receptor B6 specific IgG was purified by Human EphB6 / Eph Receptor B6 affinity chromatography.
Catalog No. 50-172-2808