Learn More
Sino Biological Recombinant Human JAM-A/F11 Receptor Protein (hFc Tag)
Supplier:  Sino Biological 10198H02H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ala 242) of human JAM-A (NP_058642.1) precursor was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4292