Learn More
Sino Biological Anti-JAM-A/F11 Receptor Antibody (APC), Mouse Monoclonal
Supplier:  Sino Biological 10198MM04A25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human JAM-A / F11R / CD321 (rh JAM-A / F11R / CD321; Catalog#10198-H08H; NP_058642.1; Met 1-Ala 242) and conjugated with APC under optimum conditions, the unreacted APC was removed.
Catalog No. 50-172-2828