Learn More
Sino Biological Anti-JAM-A/F11 Receptor Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10198RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human JAM-A extracellular domain (rhJAM-A; Catalog#10198-H08H; Met 1-Ala 242; NP_058642.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-2837