Learn More
Sino Biological Anti-Kallikrein 13 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10199MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human KLK13 / Kallikrein13 (rhKLK13; Catalog#10199-H08H; NP_056411.1; Met 1-Ile 262). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-2852