Learn More
Sino Biological Anti-Kallikrein 13 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10199RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human KLK13 / Kallikrein13 (rh KLK13; Catalog#10199-H08H; NP_056411.1; Met 1-Ile 262). KLK13 specific IgG was purified by human KLK13 affinity chromatography.
Catalog No. 50-172-2864